Please login to your online account to display your discounted pricing

MP Biomedicals Hyaluronidase, Ovine testes


Source: Ovine Testes

Manufacturer: mp biomedicals  15127480

 View more versions of this product

Catalog No. ICN15127480

  • / Each
 

Description & Specifications

Specifications

Color White
Source Ovine testes
Inhibitors Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
Concentration >1500U/mg
Molecular Weight ≈55
Product Name Hyaluronidase
Format Lyophilized
Quantity 100mg
pH 4.5 to 6
For Use With (Application) Drug dispersion and delivery
Storage Requirements -20°C