missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Human Serum, Type AB, Normal Pool
Used as a growth supplement for cell culture media and in other research applications.
Manufacturer: MP Biomedicals™ 092930949
Specifications
Grow many Human cell lines at a faster rate | |
Sterile | |
U.S. regulations | |
≤26mg/dL | |
No | |
5 to 8.3g/dL | |
0.1μm Filtration | |
Human Type AB Serum, (HBsAG), HIV-free |
100mL | |
Straw Yellow to Golden Brown | |
Liquid | |
263 to 323mOsm/kg | |
7 to 8 | |
-20°C | |
Human |