Please login to your online account to display your discounted pricing

MP Biomedicals Human Serum: Normal Serum

Hepatitis Associated Antigen (HBsAG)–free, HIV–free

$411.50 - $411.50

Specifications

Product Type Human Serum
Quantity 100mL
Source Human
View More Specs
Products
Catalog Number Mfr. No. Source Sterilization Method Sterile Price Quantity    

ICN823201

 
mp biomedicals
823201
Human 0.1μm Filtration Yes Each for $411.50
Description & Specifications

Specifications

Product Type Human Serum
Quantity 100mL
Source Human