Please login to your online account to display your discounted pricing

MP Biomedicals Human Serum: Normal Serum


Hepatitis Associated Antigen (HBsAG)–free, HIV–free

Manufacturer: mp biomedicals  823201

 View more versions of this product

Catalog No. ICN823201

  • / Each
 

Description & Specifications

Specifications

Format Liquid
For Use With (Application) Immunoassays, mammalian cell culture
Hemoglobin Concentration 1.6mg/dL
Color Clear Straw Yellow
pH 7.4
Protein Content 5.6mg/dL
Storage -20°C
Sterile Yes
Sterilization Method 0.1μm Filtration
Source Human
Quantity 100mL
Product Type Human Serum
Endotoxin Concentration 1.6EU/mL
Mycoplasma Detected No
Storage Requirements -20°C