Please login to your online account to display your discounted pricing

MP Biomedicals Human Cytokines

Manufacturer: mp biomedicals  0215134210

 View all options for this product

Catalog No. ICN15134210


Description & Specifications


Quantity 10KU
Reactivity Human, Mouse
Endotoxin Level 0.1ng/μg
Purity >97% by SDS-PAGE and HPLC analysis
Source E. coli
Sterile Yes
Storage Requirements -70°C

Suitable for supplementing cell cultures and investigating various metabolic processes. Store as detailed on label.