missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Gluten
Water insoluble protein derived from wheat
Supplier: MP Biomedicals™ 0210181510
Description
- Format: Yellow grain
- Protein Content: ∽80% ; Total Nitrogen ∽12.3
Molecular biology and Pharmaceuticals
Specifications
Molecular Biology, Pharmaceuticals | |
>100000 | |
6 to 9 | |
Wheat |
Powder | |
Gluten from Wheat | |
10000g | |
15° to 30°C |