missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Gluten

Water insoluble protein derived from wheat

$793.41 - $793.41

Specifications

Format Powder
For Use With (Application) Molecular Biology, Pharmaceuticals
Molecular Weight >100000
Name Gluten from Wheat
pH Range 6 to 9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10181510
View Documents
MP Biomedicals™
0210181510
10000g Each for $793.41
 
Description

Description

  • Format: Yellow grain
  • Protein Content: ∽80% ; Total Nitrogen ∽12.3

Molecular biology and Pharmaceuticals

Specifications

Specifications

Powder
>100000
6 to 9
15° to 30°C
Molecular Biology, Pharmaceuticals
Gluten from Wheat
Wheat