missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteriodes
Catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone
Manufacturer: MP Biomedicals™ 0215118605
Description
Cytosolic enzyme in the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells (such as erythrocytes) by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH).
- Format: White milky liquid
- Molecular Weight: 104,000 (two subunits of ∽55,000) (Lit.)
- Specific Activity: ≥250units/mg protein
Specifications
Glucose-6-phosphate Dehydrogenase | |
Liquid | |
White | |
7.8 | |
5kU | |
4°C |
Lueconostoc mesenteroides | |
≥250U/mg | |
Acyl-CoA, Metal ions | |
104000 | |
Enzyme in Pentose Phosphate pathway |