Please login to your online account to display your discounted pricing

MP Biomedicals γ-Globulin from Rabbit Plasma

Purity: ≥97%

Manufacturer: mp biomedicals  19148105

 View all options for this product

Catalog No. ICN19148105


Description & Specifications


Concentration 50mg/mL
Format Lyophilized
Formulation The product is lyophilized, in 0.9% NaCl (50mg/mL clear to turbid solution)
Name g-Globulin from Rabbit Plasma
pH Range 6.5
Quantity 5g
Source Rabbit Plasma
Storage Requirements 0°C

White, crystalline, lyophilized powder.