missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Geneclean™ Turbo Kit for PCR
Geneclean™ Turbo Kit optimized for PCR purification of DNA fragments of sizes 100 bp to 10 kb.
Supplier: MP Biomedicals™ 111103200
Description
- Ready for direct use
- Removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities
- Applications: DNA Clean-up
Specifications
Post amplification PCR reaction mixture (oil-free) | |
GeneClean™ Turbo™ Salt Solution, Wash Solution, Cartridges, Catch Tubes and Elution Solution | |
Up to 100 μL PCR Reaction Solution |
Isolation of DNA from PCR reactions, removal of primers, Nucleotides, Enzymes, Mineral Oil, salts and other impurities from DNA samples | |
50 Preps | |
Gels and Solutions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.