missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Geneclean™ Turbo Kit for PCR

Optimized for PCR purification of DNA fragments of sizes 100 bp to 10 kb.

Manufacturer:  MP Biomedicals™ 111103600

 View more versions of this product

Catalog No. MP111103600



Includes: GeneClean™ Turbo Salt Solution, Wash Solution, Cartridges, Catch Tubes and Elution Solution

Description

Description

  • Ready for direct use
  • Removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities

Applications:

DNA Clean-up

Specifications

Specifications

Isolation of DNA from PCR reactions, removal of primers, Nucleotides, Enzymes, Mineral Oil, salts and other impurities from DNA samples
Geneclean™ Turbo™ Salt solution, Wash solution, cartridges, catch tubes and Elution solution
300 Preps
Documents