missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Geneclean™ Turbo Kit

Used for purification of DNA fragments of sizes 0.1 kb to 300 kb from agarose gels, PCR reactions and other enzymatic solutions. MP Biomedicals™ Geneclean™ Turbo Kit can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.

$222.98 - $944.98

Specifications

For Use With (Application) Nucleic Acid isolation and purification
Format Geneclean™ imbedded Turbo™ SPIN column
Storage Requirements Room Temperature
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111102600 View Documents MP Biomedicals™
111102600
300 preps Each for $944.98
 
MP111102200 View Documents MP Biomedicals™
111102200
50 preps Each for $222.98
 
Description

Description

  • Process with either vacuum or centrifugation and save time with patented Turbo filter design
  • Patented GeneClean™ technology simplifies the process of purifying DNA into three easy steps: Bind, Wash, and Elute
  • Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications
Specifications

Specifications

Nucleic Acid isolation and purification
Geneclean™ imbedded Turbo™ SPIN column
Room Temperature

Research Use Only (RUO)