missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ FastPROTEIN™ BlueMatrix

Easy lysis of bacterial cells to release recombinant proteins

Manufacturer:  MP Biomedicals™ 116550500

 View more versions of this product

Catalog No. MP116550500



Includes: Matrix B, 0.1mm zirconium silicate,Tubes

Requires : FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin 2.0ml tubes, Microcentrifuge tubes (2.0ml and 1.5ml), 1x PBS or Desired Lysis Buffer

Description

Description

  • Kit or Assay Type: Protein Isolation, Mini-prep
  • Molecule to be Enriched: Proteins
  • Sample Type: Bacteria, Gram +/-
  • Tube Size: 2mL
  • Matrix Composition: Matrix B, 0.1mm zirconium silicate

Specifications

Specifications

Blue matrix kit
Rapid isolation and purification of Protein from bacterial cultures
15° to 30°C
100 Preps
Matrix B, 0.1mm Zirconium Silicate, tubes
Documents