missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ EMM, Powdered Media

Defined medium for the growth of S. pombe

Manufacturer:  MP Biomedicals™ 114110032

 View more versions of this product

Catalog No. MP114110032



Description

Description

Formulation

  • Contents/liter: 3g phthalic Acid K+, 2.2g Na2HPO4, 5g NH4Cl, 20g dextrose
  • Salts: 1.05g MgCl2·6H2O, 4.7mg CaCl2·2H2O, 1g KCl, 0.04g Na2SO4
  • Vitamins: 1mg pantothenic acid, 10mg nicotinic acid, 10mg myo-inositol, 1mg biotin
  • Minerals: 0.5mg boric acid, 0.4mg MnSO4, 0.4mg ZnSO4·7H2O, 0.2mg FeCl2·6H2O, 40μg molybdic acid, 0.1mg KI, 40μg CuSO4·5H20, 1mg citric acid

Applications:

Yeast cell culture, Life sciences

Specifications

Specifications

Certified Thiamine (Vitamin B1) free
Powder
1000g
EMM
Molecular biology
phthalic Acid K+ 3g/L, Na2HPO4 2.2g/L, NH4Cl 5g/L, Dextrose 20g/L, MgCl2.6H2O 1.05g/L, CaCl2.2H2O 4.7mg/L, KCl 1g/L, Na2SO4 0.04g/L, Pantothenic Acid 1mg/L, Nicotinic Acid 10mg/L, myo-inositol 10mg/L, Biotin 1mg/L, Boric Acid 0.5mg/L, MnSO4 0.4mg/L, ZnSO4.7H2O 0.4mg/L, FeCl2.6H2O 0.2mg/L, Molybdic Acid 40μg/L, KI 0.1mg/L, CuSO4.5H2O 40μg/L, Citric Acid 1mg/L
4°C
For growth of fission Yeast S. Pombe
Documents