Please login to your online account to display your discounted pricing

MP Biomedicals Elastase, Highly Pure, Salt Free


>95u/mg protein, Porcine Pancreas, Clear, to White Fluffy Solid

Manufacturer: mp biomedicals  10061905

 View more versions of this product

Catalog No. ICN10061905

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized Powder
Source Porcine pancreas
For Use With (Application) Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
Color Clear to White
Concentration 2X
Storage Requirements 4°C
Quantity 5mg
Inhibitors DFP, elastinal, alpha-2-macroglobulin
Product Name Elastase
Molecular Weight 25500