Please login to your online account to display your discounted pricing

MP Biomedicals Elastase, Highly Pure, Salt Free

>95u/mg protein, Porcine Pancreas, Clear, to White Fluffy Solid

$219.56 - $219.56

Specifications

Color Clear to White
Concentration 2X
Product Name Elastase
Format Lyophilized Powder
For Use With (Application) Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10061905

 
mp biomedicals
10061905
5mg Each for $219.56
Description & Specifications

Specifications

Color Clear to White
Concentration 2X
Product Name Elastase
Format Lyophilized Powder
For Use With (Application) Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
Inhibitors DFP, elastinal, alpha-2-macroglobulin
Molecular Weight 25500
Source Porcine pancreas
Storage Requirements 4°C

Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.

  • Inhibitors: DFP, elastinal, α-2-macroglobulin
  • Synonyms: Elastase from hog pancreas; Pancreatopeptidase
  • Format: Solid
  • MDL Number MFCD00130998
  • Source: Porcine Pancreas
  • Enzyme Commission Number: E.C.3.4.21.36