missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Dispase
From Bacillus polymyxa
Manufacturer: MP Biomedicals™ 0219502205
Description
Microbial metalloenzyme used in tissue dissociation (primary cell cultures) and subcultivation (secondary cell cultures). Extremely stable. Activity: 6U/mg. Unit Definition: One unit is the amount of enzyme which liberates 1.0μmole of Tyr per minute at 37°C, pH 7.5. MW: 35.9kDa. Lyophilized.
Ref.: Akimoto, Y., et al., J. Histochem. Cytochem., 40: 1129 (1992).
Specifications
Dispase | |
Tan | |
Bacillus polymyxa | |
35.9 | |
Protein sequencing |
Lyophilized Powder | |
EDTA, EGTA, Hg2+, other heavy Metals | |
6 to 8.5 | |
5U | |
4°C |