missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Dextrose
Monohydrate
Manufacturer: MP Biomedicals™ 0290559425
Description
Dextrose monohydrate is a common natural sugar involved in processes such as energy production, glycosylation and formation of glycans that provide structure to cells. It is involved in a detrimental process in cells called glycation. It is used as a supplement for cell culture and in numerous cellular processes.
Formula: C6H12O6•H2O
Formula Weight: 198.18
CAS No.: 14431-43-7
Specifications
Animal Diet | |
Used as a supplement for cell culture and in numerous cellular processes | |
15°C to 30°C |
Powder | |
11.33 kg |