Please login to your online account to display your discounted pricing

MP Biomedicals D-3-Hydroxybutyrate Dehydrogenase from Pseudomonas species

Manufacturer: mp biomedicals  0215348480

 View all options for this product

Catalog No. ICN15348480


Description & Specifications


Product Name D-3-Hydroxybutyrate Dehydrogenase
Format Lyophilized Powder
Concentration 100U/mg
Inhibitors PCMB, MIA, IAA, Ag+, Hg+, SDS, DAC
Source Pseudomonas species
pH 8.3
Molecular Weight ≈130000
Quantity 100U
Storage Requirements -20°C