Please login to your online account to display your discounted pricing

MP Biomedicals D-3-Hydroxybutyrate Dehydrogenase from Pseudomonas species

$2,377.78 - $7,207.10

Specifications

Product Name D-3-Hydroxybutyrate Dehydrogenase
Inhibitors PCMB, MIA, IAA, Ag+, Hg+, SDS, DAC
Molecular Weight ≈130000
pH 8.3
Source Pseudomonas species
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15348425

 
mp biomedicals
0215348425
25U Each for $2,377.78

ICN15348480

 
mp biomedicals
0215348480
100U Each for $7,207.10
Description & Specifications

Specifications

Product Name D-3-Hydroxybutyrate Dehydrogenase
Inhibitors PCMB, MIA, IAA, Ag+, Hg+, SDS, DAC
Molecular Weight ≈130000
pH 8.3
Source Pseudomonas species
Storage Requirements -20°C