Please login to your online account to display your discounted pricing

MP Biomedicals CSM, Powdered Media

Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

$160.58 - $1,203.97

Specifications

Certifications/Compliance Molecular biology Certified™
Format Powder
For Use With (Application) For growth and maintenance of M13 Phage or other filamentous Phage
Product Type CSM
Grade Molecular biology
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MP114500012

 
mp biomedicals
114500012
10g Each for $160.58

MP114500022

 
mp biomedicals
114500022
100g Each for $1,203.97
Description & Specifications

Specifications

Certifications/Compliance Molecular biology Certified™
Format Powder
For Use With (Application) For growth and maintenance of M13 Phage or other filamentous Phage
Product Type CSM
Grade Molecular biology
Media Format Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, Uracil 20mg/L, L-Tyrosine 50mg/L, L-Valine 140mg/L, Total 790mg/L
Shelf Life 4 Years
Storage Requirements 15-30°C

Formulation

  • Add 0.79g of CSM to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae

Applications:

Yeast cell culture, Gene expression, Transcription regulation

Safety and Handling

4 Years