Please login to your online account to display your discounted pricing

MP Biomedicals CSM-LEU-URA , Powdered Media

Dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

$160.58 - $1,203.97

Specifications

Certifications/Compliance Molecular biology Certified™
Format Powder
For Use With (Application) For growth of Saccharomyces cerevisiae
Product Type CSM-LEU
Media Format Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MP114520222

 
mp biomedicals
114520222
100g Each for $1,203.97

MP114520212

 
mp biomedicals
114520212
10g Each for $160.58
Description & Specifications

Specifications

Certifications/Compliance Molecular biology Certified™
Format Powder
For Use With (Application) For growth of Saccharomyces cerevisiae
Product Type CSM-LEU
Media Format Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L
Shelf Life 4 Years
Storage Requirements 15-30°C

  • Add 0.67g of CSM-Leu-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium

Applications:

Yeast cell culture, Gene expression, Transcription regulation