missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ CSM-HIS-LEU-TRP, Powdered Media
Dropout supplement for S. cereviseae
$188.00 - $1029.17
Specifications
Certifications/Compliance | Molecular biology Certified™ |
---|---|
Content And Storage | 15°C to 30°C |
Form | Powder |
Product Type | Yeast Media |
Formulation | Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
MP114530112
|
MP Biomedicals™
114530112 |
10 g |
Each for $188.00
|
|
|||||
MP114530122
|
MP Biomedicals™
114530122 |
100 g |
Each for $1,029.17
|
|
|||||
Description
Formulation
- Add 0.62g of CSM-His-Leu-Trp to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined triple drop-out medium
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |