missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ CSM-HIS-LEU, Powdered Media

Dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

Manufacturer:  MP Biomedicals™ 114520422

 View more versions of this product

Catalog No. MP114520422



Description

Description

Formulation

  • Add 0.67g of CSM-His-Leu to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium

Applications:

Yeast cell culture, Gene expression, Transcription regulation

Specifications

Specifications

Molecular biology Certified™
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
15-30°C
For growth of Saccharomyces cerevisiae
Powder
100g
CSM-HIS-LEU
Documents