missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Collagenase, Type 4

From Clostridium histolyticum

$60.30 - $321.55

Specifications

Color Brown
Concentration >160U/mg
Product Type Collagenase Type 4
Format Lyophilized Powder
For Use With (Application) For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19511025
View Documents
MP Biomedicals™
0219511025
25mg Each for $60.30
 
ICN19511080
View Documents
MP Biomedicals™
0219511080
100mg Each for $102.92
 
ICN19511090
View Documents
MP Biomedicals™
0219511090
500mg Each for $321.55
 
Description

Description

Pancreatic Islet Isolation Grade. Suggested for pancreatic islet isolation. Prepared to contain lower tryptic activity levels. Activity: ≥160U per mg solid. Unit Definition: One unit liberates peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Dialyzed, lyophilized.

Specifications

Specifications

Brown
Collagenase Type 4
For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain
Clostridium histolyticum
>160U/mg
Lyophilized Powder
Cysteine, EDTA or o-phenanthroline, alpha2-macroglobulin - a large plasma glycoprotein, alpha2-macroglobulin, alpha1-antitrypsin
4°C