missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Collagenase, Type 4
From Clostridium histolyticum
Supplier: MP Biomedicals™ 0219511025
Description
Pancreatic Islet Isolation Grade. Suggested for pancreatic islet isolation. Prepared to contain lower tryptic activity levels. Activity: ≥160U per mg solid. Unit Definition: One unit liberates peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Dialyzed, lyophilized.
Specifications
>160 U/mg | |
For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain | |
Clostridium histolyticum | |
Collagenase Type 4 | |
Lyophilized Powder |
Brown | |
4°C | |
25 mg | |
Cysteine, EDTA or o-phenanthroline, alpha2-macroglobulin - a large plasma glycoprotein, alpha2-macroglobulin, alpha1-antitrypsin |