missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ CIRCLEGROW™, Powder

Superior plasmid yield through enhanced cell growth

Manufacturer:  MP Biomedicals™ 113000185

 View more versions of this product

Catalog No. MP113000185



Description

Description

Perfect for overnight or weekend cultures, CIRCLEGROW Medium is a super-rich bacterial growth medium that produces up to three times more plasmid DNA than traditional growth medium. CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death during the log phase of growth.

  • Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed
  • Reduce hands-on work and preparation time
  • Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
  • Obtain high yields of plasmid DNA from smaller culture volumes, and confidently leave cultures overnight or over the weekend
  • Capsules are the most convenient form of this unique growth media
  • Pre-mixed CIRCLEGROW is also available in single-use pouches, capsules and sterile liquid
Formulation
  • MP113000165: Includes ten 20 g pouches. Add contents of one pouch to 500mL of purified water.
  • MP113000175: Includes ten 40g pouches. Add contents of one pouch to 1L of purified water.
  • MP113000185: Includes one hundred 20g pouches. Add contents of one pouch to 500mL of purified water.

Specifications

Specifications

Sterile
Culture Medium
15° to 30°C
6.9
100 x 0.5L
Documents