missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ ChromatoPur™ Bovine IgG, ImmunoAssay

Bovine IgG is suitable as a standard and coating reagent for ELISA

Manufacturer:  MP Biomedicals™ 08641403

 View more versions of this product

Catalog No. MP08641403



Description

Description

Purified using gentle chromatographic extraction.

Physical Characteristics

  • Appearance: Off-white powder
  • Purity (SDS-PAGE): ≥95% (IgG w/w Total Protein
  • Total Protein (anhydrouse): ≥96%
  • pH (4%): 4.0 to 5.0
  • Moisture: ≤10%
  • Sodium: ≤1.5% w/w
  • Chloride: ≤2.4% w/w
\
Certifications: ISO, cGMP, EDQM

Specifications

Specifications

ISO, cGMP, EDQM
100g
4°C. Re-test date: Five years from the date of manufacture.
Documents