missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ ChromatoPur™ Bovine IgG, ImmunoAssay
Bovine IgG is suitable as a standard and coating reagent for ELISA
Supplier: MP Biomedicals™ 08641402
Description
Purified using gentle chromatographic extraction.
Physical Characteristics
- Appearance: Off-white powder
- Purity (SDS-PAGE): ≥95% (IgG w/w Total Protein
- Total Protein (anhydrouse): ≥96%
- pH (4%): 4.0 to 5.0
- Moisture: ≤10%
- Sodium: ≤1.5% w/w
- Chloride: ≤2.4% w/w
Certifications: ISO, cGMP, EDQM
Specifications
10 g | |
4°C. Re-test date: Five years from the date of manufacture. |