missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Casein Hydrolysate

5% Sterile Solution

Manufacturer:  MP Biomedicals™ 0210486490

Catalog No. ICN10486490



Description

Description

  • Format: Liquid
  • Vitamin Free
  • Sterile
  • Used as nutritional source in microbial media

Specifications

Specifications

Liquid
Casein Hydrolysate Enzymatic
500mL
15° to 30°C
SDS
Documents
Product Certifications
Promotions

Promotions