missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Casein Hydrolysate

5% Sterile Solution

Manufacturer:  MP Biomedicals™ 10486480

 View more versions of this product

Catalog No. ICN10486480



Description

Description

  • Format: Liquid
  • Vitamin Free
  • Sterile
  • Used as nutritional source in microbial media

Specifications

Specifications

Liquid
Casein Hydrolysate Enzymatic
100mL
15° to 30°C
Documents