missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Basal Medium Eagle Vitamin Concentrate, 100X, Modified

For use with Mammalian Cell Culture

$113.46 - $113.46

Specifications

pH 5.18
Solution Type BME
Sterility Sterile
Storage -20°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN1600449 View Documents MP Biomedicals™
1600449
100mL Each for $113.46
Specifications

Specifications

5.18
BME
Sterile
-20°C