Please login to your online account to display your discounted pricing

MP Biomedicals Bacteriological Agar


Specially purified for use in preparing solid culture media for microbiological and bacterilogical applications

Manufacturer: mp biomedicals  19402191

 View more versions of this product

Catalog No. ICN19402191

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 15-30°C
Grade Bacteriological
Format Powder
Includes 2U of Agarose and Agaropectin
Product Type Agar
Quantity 1kg
For Use With (Application) Used for preparing solid culture media for microbiological and bacteriological application, antistaling agent, gelling agent
Media Format Agarose 70% and Agaropectin 30%