missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Bacteriological Agar

Specially purified for use in preparing solid culture media for microbiological and bacterilogical applications

Manufacturer:  MP Biomedicals 19402183

 View more versions of this product

Catalog No. ICN19402183


 

Specifications Description & Specifications

Specifications

Used for preparing solid culture media for microbiological and bacteriological application, antistaling agent, gelling agent
Powder
Agarose 70% and Agaropectin 30%
250g
Bacteriological
2U of Agarose and Agaropectin
Agar
15-30°C