Please login to your online account to display your discounted pricing

MP Biomedicals Bacteriological Agar

Specially purified for use in preparing solid culture media for microbiological and bacterilogical applications

$259.48 - $852.92

Specifications

Format Powder
For Use With (Application) Used for preparing solid culture media for microbiological and bacteriological application, antistaling agent, gelling agent
Product Type Agar
Grade Bacteriological
Includes 2U of Agarose and Agaropectin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19402183

 
mp biomedicals
19402183
250g Each for $259.48

ICN19402191

 
mp biomedicals
19402191
1kg Each for $852.92
Description & Specifications

Specifications

Format Powder
For Use With (Application) Used for preparing solid culture media for microbiological and bacteriological application, antistaling agent, gelling agent
Product Type Agar
Grade Bacteriological
Includes 2U of Agarose and Agaropectin
Media Format Agarose 70% and Agaropectin 30%
Storage Requirements 15-30°C

Naturally occurring impurities have been reduced to a minimum

  • CAS: 9002-18-0
  • EC No. 232-658-1