Please login to your online account to display your discounted pricing

MP Biomedicals Antibiotics: Nystatin


Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis

Manufacturer: mp biomedicals  19453405

 View more versions of this product

Catalog No. ICN19453405

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Solution Type Nystatin
Storage Requirements 4°C
Format Powder
Quantity 5mU