α-Amylase from Porcine pancreas, MP Biomedicals™
α-Amylase from Porcine pancreas ≥700 u/mg protein, Opaque, White Suspension
Manufacturer:  Mp Biomedicals 19123910
Catalog No. ICN19123910
Description
Description
  • It is used to hydrolyze α bonds of α-linked polysaccharides, such as starch and glycogen.
  • It has been used in various plant studies, such as metabolism studies in Arabidopsis.
  • It is isolated from porcine pancreas is a glycoprotein.
  • It is a single polypeptide chain of ∽475 residues containing two SH groups and four disulfide bridges and a tightly bound Ca2+ necessary for stability. Chloride ions are necessary for activity and stability.
Specifications
Specifications
Opaque White,Opaque White
5.5 to 8.0 (activity range), 7 (optimum)
Porcine pancreas
Molecular biology
MFCD00081319
Suspension
alpha-Amylase
Suspension
10mg
4°C
β-N-acetylglucosaminidase porcine placenta, 4glucan-4-glucanohydrolase, PPA, al1
Documents