missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ alpha-Amylase from Bacillus subtilis
Liquefying diastase supplied as dry, free-flowing powder with high uniformity and storage stability
Manufacturer: MP Biomedicals™ 0210044780
Description
Deactivation temperature: 90 to 100°CSpecifications
alpha-Amylase | |
Powder | |
60 to 80°C | |
100g | |
4°C |
Bacillus subtilis | |
Off-White to Tan | |
5 to 7 | |
Carbohydrate hydrolysis |