Please login to your online account to display your discounted pricing

MP Biomedicals AIN-76 Vitamin Mixture

Powdered

$242.84 - $242.84

Specifications

Components Thiamine Hydrochloride 0.6g, Riboflavin 0.6g, Pyridoxine Hydrochloride 0.7g, Nicotinic Acid 3g, D-Calcium Pantothenate 1.6g, Folic Acid 0.2g, D-Biotin 0.02g, Cyanocobalamin (Vitamin B12) 0.001g, Retinyl Palmitate (Vitamin A) Pre-mix (250000 IU/g) 1.6g, DL-a-Tocopherol Acetate (250 IU/g)20g, Cholecalciferol (Vitamin D3, 400000 IU/g) 0.25g, Menaquinone (Vitamin K2) 0.005g, Sucrose, finely powdered 972.9g
Product Type AIN-76 Vitamin mixture
Quantity 1000g
For Use With (Application) AIN-76 Vitamin mixture is a basic component of animal diet for maintaining Mice and Rats colonies in the research lab
Species Rat
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

ICN90545401

 
mp biomedicals
90545401
Powder Each for $242.84
Description & Specifications

Specifications

Components Thiamine Hydrochloride 0.6g, Riboflavin 0.6g, Pyridoxine Hydrochloride 0.7g, Nicotinic Acid 3g, D-Calcium Pantothenate 1.6g, Folic Acid 0.2g, D-Biotin 0.02g, Cyanocobalamin (Vitamin B12) 0.001g, Retinyl Palmitate (Vitamin A) Pre-mix (250000 IU/g) 1.6g, DL-a-Tocopherol Acetate (250 IU/g)20g, Cholecalciferol (Vitamin D3, 400000 IU/g) 0.25g, Menaquinone (Vitamin K2) 0.005g, Sucrose, finely powdered 972.9g
Product Type AIN-76 Vitamin mixture
Quantity 1000g
For Use With (Application) AIN-76 Vitamin mixture is a basic component of animal diet for maintaining Mice and Rats colonies in the research lab
Species Rat
Storage Requirements 15 to 30°C