Please login to your online account to display your discounted pricing

MP Biomedicals Agar, 80-100 Mesh


Cell Culture Reagent

Manufacturer: mp biomedicals  19461591

 View more versions of this product

Catalog No. ICN19461591

  • / Each
 

Description & Specifications

Specifications

Format Powder
Storage Requirements 15° to 30°C
Quantity 1kg
pH 7 to 7.6
Product Type Agar