Please login to your online account to display your discounted pricing

MP Biomedicals Agar, 80-100 Mesh


Cell Culture Reagent

Manufacturer: mp biomedicals  19461583

 View more versions of this product

Catalog No. ICN19461583

  • / Each
 

Description & Specifications

Specifications

Product Type Agar
Format Powder
pH 7 to 7.6
Storage Requirements 15° to 30°C
Quantity 250g