Please login to your online account to display your discounted pricing

MP Biomedicals Agar, 80-100 Mesh

Cell Culture Reagent

$68.29 - $1,260.29

Specifications

Format Powder
Product Type Agar
pH 7 to 7.6
Storage Requirements 15° to 30°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19461580

 
mp biomedicals
19461580
100g Each for $68.29

ICN19461583

 
mp biomedicals
19461583
250g Each for $136.59

ICN19461591

 
mp biomedicals
19461591
1kg Each for $320.76

ICN19461590

 
mp biomedicals
19461590
500g Each for $190.31

ICN19461594

 
mp biomedicals
19461594
5kg Each for $1,260.29
Description & Specifications

Specifications

Format Powder
Product Type Agar
pH 7 to 7.6
Storage Requirements 15° to 30°C

Powder