Please login to your online account to display your discounted pricing

Moxonidine hydrochloride, Tocris

I1 receptor and α2-adrenoceptor agonist (I1 > α2)

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Moxonidine hydrochloride
Assay Percent Range >98%
Molecular Formula C9H12ClN5O·HCl
Formula Weight 278.14
Synonym 4-Chloro-N-(4, 5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

228250

 
r & d systems
228250
50mg Each for $746.48

228210

 
r & d systems
228210
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 278.14
Quantity 50mg
Synonym 4-Chloro-N-(4, 5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine hydrochloride
CAS 75536-04-8
Recommended Storage Desiccate at +4°C
Chemical Name or Material Moxonidine hydrochloride
Molecular Formula C9H12ClN5O·HCl
Assay Percent Range >98%
Safety and Handling

Desiccate at +4°C