Learn More
Moxifloxacin hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J66626ME
Description
Moxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMoxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.
Solubility
Sparingly soluble in water. Soluble in DMSO
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Moxifloxacin Hydrochloride | |
Powder | |
C21H25ClFN3O4 | |
Odorless | |
Sparingly soluble in water; Soluble in DMSO | |
COC1=C2C(=CC(=C1N3CC4CCCNC4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl | |
437.896 | |
CHEBI:7008 | |
97% | |
500 mg | |
636°C |
Moxifloxacin hydrochloride | |
186826-86-8 | |
MFCD00949117 | |
8377447 | |
IDIIJJHBXUESQI-DFIJPDEKSA-N | |
7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride | |
101526 | |
437.89 | |
Yellow | |
>110°C (230°F) |
Chemical Identifiers
186826-86-8 | |
437.896 | |
IDIIJJHBXUESQI-DFIJPDEKSA-N | |
CHEBI:7008 | |
COC1=C2C(=CC(=C1N3CC4CCCNC4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl |
C21H25ClFN3O4 | |
MFCD00949117 | |
101526 | |
7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride |
Safety and Handling
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
RTECSNumber : VB1983750
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only