missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Moxalactam sodium salt, 50 mg/mL in distilled water, sterile-filtered, Thermo Scientific™
$126.92 - $351.03
Chemical Identifiers
CAS | 64953-12-4 |
---|---|
Molecular Formula | C20H18N6Na2O9S |
Molecular Weight (g/mol) | 564.437 |
MDL Number | MFCD03427565 |
InChI Key | GRIXGZQULWMCLU-GDUWRUPCSA-L |
PubChem CID | 12856838 |
IUPAC Name | disodium;(6R,7R)-7-[[(2R)-2-carboxylato-2-(4-hydroxyphenyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
SMILES | CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C(C4=CC=C(C=C4)O)C(=O)[O-])OC)OC2)C(=O)[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67056XF
|
Thermo Scientific Chemicals
J67056XF |
1 mL |
Each for $126.92
|
|
|||||
AAJ670568EQ
|
Thermo Scientific Chemicals
J670568EQ |
5 x 1 mL |
Pack for $351.03
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
64953-12-4 | |
564.437 | |
GRIXGZQULWMCLU-GDUWRUPCSA-L | |
disodium;(6R,7R)-7-[[(2R)-2-carboxylato-2-(4-hydroxyphenyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
C20H18N6Na2O9S | |
MFCD03427565 | |
12856838 | |
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C(C4=CC=C(C=C4)O)C(=O)[O-])OC)OC2)C(=O)[O-].[Na+].[Na+] |
Specifications
Moxalactam Sodium Salt | |
64953-12-4 | |
MFCD03427565 | |
GRIXGZQULWMCLU-GDUWRUPCSA-L | |
disodium;(6R,7R)-7-[[(2R)-2-carboxylato-2-(4-hydroxyphenyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
12856838 | |
Moxalactam sodium salt |
Liquid | |
C20H18N6Na2O9S | |
Plastic Bottle | |
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C(C4=CC=C(C=C4)O)C(=O)[O-])OC)OC2)C(=O)[O-].[Na+].[Na+] | |
564.437 | |
1 mL |
Safety and Handling
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 265-288-4
TSCA : No
Recommended Storage : Store at -20°C