Please login to your online account to display your discounted pricing

Mouse VEGF Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSMMV00

 View more versions of this product

Catalog No. MMV00

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
For Use With (Application) ELISA
Gene ID (Entrez) 22339
Packaging Quantity 2 Plates
Quantity 1 Kit
Sample Type Cell Culture Supernates, Tissue Homogenates, Serum, EDTA Plasma
Sample Volume Cell Culture Supernates (50μL), Tissue Homogenates (50μL), Serum (10μL), EDTA Plasma (10μL)
Species Mouse