missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Mouse TPO (Thrombopoietin) (Carrier-Free) Recombinant Protein, eBioscience™
Recombinant Protein
Supplier: Invitrogen™ 34868682
Description
Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa. Description: Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa. Applications Reported: Recombinant human TPO is biologically active and can promote proliferation of human MO7e cells in culture. Applications Tested: The ED50 of this protein, as measured by proliferation of human MO7e cells, is less than or equal to 1.0ng/mL, which corresponds to a specific activity of at least 1.0 x 10e6 Units/mg. Source: E. coli (amino acids Glu22-Thr195; accession # NP_001166976.1). Bioactivity: The ED50 of this protein, as measured by proliferation of human MO7e cells, is less than or equal to 1.0ng/mL, which corresponds to a specific activity of at least 1.0 x 10e6 Units/mg. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE and HPLC. Molecular Weight: 18.7 kDa. Storage and handling: For best recovery, quick spin vial prior to opening. Use in sterile environment. Reconstitute in water too a concentration of less than or equal to 0.5mg/mL. This protein dissolves slowly. Do not vortex. Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier-protein (e.g., 0.1% BSA) at less than or equal to -20°C.Specifications
P40226 | |
0.01M tris with no preservative; pH 8.0 | |
18.7 | |
8.0 | |
100μg | |
-20°C | |
E. coli (amino acids Glu22-Thr195; Accession # NP_001166976.1) | |
Mouse Thrombopoietin (TPO) Carrier-Free | |
The ED50 of this protein, as measured by proliferation of human MO7e cells, is less than or equal to 1.0ng/mL, which corresponds to a specific activity of at least 1.0 x 106 Units/mg. | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
Functional Assay | |
21832 | |
Mouse Thrombopoietin (TPO) Carrier-Free | |
SDS-PAGE and HPLC | |
E. coli | |
RUO | |
Less than 0.1ng/μg cytokine as determined by the LAL assay | |
Thpo | |
Recombinant Protein, Carrier-Free | |
Unconjugated | |
E. coli | |
Lyophilized | |
Affinity chromatography |