Please login to your online account to display your discounted pricing

eBioscience™ Mouse TNF alpha, Recombinant Protein, Carrier-Free

Mouse Recombinant Protein

$274.05 - $2,275.05

Specifications

Name TNF alpha
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

509204

 
ebioscience™
LS34832163
10μg Each for $274.05

509205

 
ebioscience™
LS34832182
100μg Each for $752.55

509206

 
ebioscience™
LS34832185
500μg Each for $2,275.05
Description & Specifications

Specifications

Name TNF alpha
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight The protein is not methionylated at the N-terminal and has a predicted molecular mass of 17,258. The DTT-reduced and non-reduced protein migrate as 17kDa polypeptides on SDS-PAGE.
Storage Requirements Store at less than or equal to -70°C.

Mouse TNF alpha is a 17 kDa factor produced by macrophages, monocytes, neutrophils, CD4+ T cells and NK cells. A 26 kDa form of TNF alpha is expressed as a membrane-bound molecule. TNF alpha is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF alpha have been observed.