missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Mouse TNF-alpha Platinum ELISA Kit

For the quantitative detection of mouse TNF-alpha.

Manufacturer:  Invitrogen™ LSBMS6073

 View more versions of this product

Catalog No. 501125453



Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to mouse TNF-alpha
Biotin-Conjugate anti-mouse TNF-alpha polyclonal antibody
Streptavidin-HRP
Mouse TNF-alpha Standard lyophilized, 2ng/mL upon reconstitution
Calibrator Diluent
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

P06804
Biotin
21926
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 tests
3.7pg/mL
50μL
Mouse
1 hr 20 min
TNF alpha
Cachectin, Tumor necrosis factor alpha, DIF
6.5%
31.3-2,000 pg/mL
ELISA
Colorimetric Microplate Reader
ELISA
RUO
Cell culture supernatant, serum, plasma (EDTA, citrate)
Plasma (50μL), Serum (50μL), Supernatants (50μL)
2-8°C
3 hr 30 min
Tnf
5.7%
HRP
Documents