missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Mouse TGF-beta1 Platinum ELISA Kit

For the quantitative detection of mouse TGF-beta1.

Manufacturer:  Invitrogen™ LSBMS6084FIVE

 View more versions of this product

Catalog No. 501125458



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse TGF-beta1
Biotin-Conjugate anti-mouse TGF-beta1 polyclonal antibody
Streptavidin-HRP
Mouse TGF-beta1 Standard lyophilized, 4ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
]Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

P04202
Biotin
21803
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
5 x 96 tests
12pg/mL
10 to 40μL (pretreated)
Mouse
1 hr 20 min
TGF beta 1
Camurati-Engelmann disease, DPD1
7.9%
31.25-2,000 pg/mL
ELISA
Colorimetric Microplate Reader
ELISA
RUO
Cell culture supernatant, serum, plasma (EDTA, citrate)
Plasma (20μL), Serum (20μL), Supernatants (20μL)
2-8°C
4 hr
Tgfb1
5.8%
HRP
Documents